Assay Details
| Gene Symbol: Adcy9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291910.1 | NP_001278839.1 | 5 - 6 | 2528 | 82 | |
| NM_009624.3 | NP_033754.2 | 4 - 5 | 2751 | 82 | ||
| GenBank mRNA | AK156940.1 | - | 5 - 6 | 2530 | 82 | |
| U30602.1 | - | 4 - 5 | 2058 | 82 | ||
| Z50190.1 | - | 3 - 4 | 2027 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adcy9
Genomic Map