Assay Details
| Gene Symbol: Adh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009626.4 | NP_033756.2 | 3 - 4 | 381 | 92 | |
| GenBank mRNA | AF331802.1 | - | 3 - 4 | 379 | 92 | |
| AF331803.1 | - | 3 - 4 | 379 | 92 | ||
| AK009588.2 | - | 3 - 4 | 348 | 92 | ||
| AK009600.1 | - | 3 - 4 | 394 | 92 | ||
| BC047267.1 | - | 3 - 4 | 310 | 6310424 | 92 | |
| BQ560660.1 | - | 3 - 4 | 381 | 92 | ||
| U20257.1 | - | 3 - 4 | 272 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adh7
Genomic Map