Assay Details
| Gene Symbol: Adh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009626.4 | NP_033756.2 | 5 - 6 | 680 | 68 | |
| GenBank mRNA | AF331802.1 | - | 5 - 6 | 678 | 68 | |
| AF331803.1 | - | 5 - 6 | 678 | 68 | ||
| AK009588.2 | - | 5 - 6 | 647 | 68 | ||
| AK009600.1 | - | 5 - 6 | 693 | 68 | ||
| AK144795.1 | - | 631 | 68 | |||
| BC047267.1 | - | 5 - 6 | 609 | 6310424 | 68 | |
| U20257.1 | - | 5 - 6 | 571 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adh7
Genomic Map