Assay Details
Gene Symbol: Angpt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009641.1 | NP_033771.1 | 5 - 6 | 970 | 85 | |
GenBank mRNA | AF113707.1 | - | 5 - 6 | 970 | 85 | |
AK132988.1 | - | 1095 | 85 | |||
AK154379.1 | - | 1095 | 85 | |||
BC129965.1 | - | 5 - 6 | 970 | 40106226 | 85 | |
BC129966.1 | - | 5 - 6 | 970 | 40106227 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
