Assay Details
| Gene Symbol: Aicda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009645.2 | NP_033775.1 | 1 - 2 | 95 | 119 | |
| GenBank mRNA | AF132979.1 | - | 1 - 2 | 94 | 119 | |
| CJ161825.1 | - | 1 - 2 | 104 | 119 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF132979.1,NM_009645.2 transcripts.
Set Membership: |
|

Aicda
Genomic Map