Assay Details
| Gene Symbol: Aes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276288.1 | NP_001263217.1 | 5 - 6 | 502 | 71 | |
| NM_010347.4 | NP_034477.1 | 5 - 6 | 502 | 71 | ||
| NR_074087.1 | 5 - 6 | 537 | 71 | |||
| GenBank mRNA | AK131845.1 | - | 5 - 6 | 379 | 71 | |
| AK131994.1 | - | 5 - 6 | 450 | 71 | ||
| AK149015.1 | - | 2 - 3 | 1170 | 71 | ||
| AK158714.1 | - | 5 - 6 | 387 | 71 | ||
| BC132217.1 | - | 5 - 6 | 390 | 40130494 | 71 | |
| BC132219.1 | - | 5 - 6 | 390 | 40130496 | 71 | |
| BC145299.1 | - | 5 - 6 | 390 | 9053853 | 71 | |
| BE685863.1 | - | 5 - 6 | 329 | 71 | ||
| BY713872.1 | - | 5 - 6 | 503 | 71 | ||
| L12140.1 | - | 5 - 6 | 343 | 71 | ||
| X73359.1 | - | 4 - 5 | 453 | 71 | ||
| X73361.1 | - | 5 - 6 | 402 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aes
Genomic Map