Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900021656.mm.1 |
NM_022026.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021656.mm.1 |
NR_073483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021656.mm.1 |
TC0900002604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021657.mm.1 |
NM_022026.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021657.mm.1 |
NR_073483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021657.mm.1 |
TC0900002604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021660.mm.1 |
NM_022026.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021660.mm.1 |
NR_073483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021660.mm.1 |
TC0900002604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021659.mm.1 |
NM_022026.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021659.mm.1 |
NR_073483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021659.mm.1 |
TC0900002604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021654.mm.1 |
NM_001271843.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021654.mm.1 |
NM_022026.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021654.mm.1 |
NR_073483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021654.mm.1 |
TC0900002604.mm.1 |
MTA TC |