Assay Details
Gene Symbol: Aqp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271843.1 | NP_001258772.1 | 3 - 4 | 579 | 78 | |
NM_022026.3 | NP_071309.1 | 3 - 4 | 579 | 78 | ||
NR_073483.1 | 3 - 4 | 579 | 78 | |||
GenBank mRNA | AB037180.1 | - | 3 - 4 | 376 | 78 | |
AK050042.1 | - | 3 - 4 | 579 | 78 | ||
AK154151.1 | - | 3 - 4 | 583 | 78 | ||
BC024105.1 | - | 3 - 4 | 550 | 5096952 | 78 | |
CT010174.1 | - | 3 - 4 | 373 | 78 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK050325.1 | - | 1635 | Chr. - - [-] on Build | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
