Assay Details
Gene Symbol: Krt20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023256.1 | NP_075745.1 | 5 - 6 | 995 | 78 | |
GenBank mRNA | AF473907.1 | - | 5 - 6 | 972 | 78 | |
AK018567.1 | - | 5 - 6 | 995 | 78 | ||
BC038900.1 | - | 1 - 2 | 82 | 78 | ||
BC057172.1 | - | 5 - 6 | 978 | 4989154 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
