Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 2 - 3 | 235 | 83 | |
| GenBank mRNA | AF172276.2 | - | 2 - 3 | 311 | 83 | |
| AK054065.1 | - | 2 - 3 | 194 | 83 | ||
| AK143048.1 | - | 2 - 3 | 215 | 83 | ||
| AK149434.1 | - | 2 - 3 | 196 | 83 | ||
| AY347569.1 | - | 2 - 3 | 112 | 83 | ||
| AY347570.1 | - | 2 - 3 | 112 | 83 | ||
| BC024046.1 | - | 2 - 3 | 185 | 83 | ||
| BC049853.1 | - | 2 - 3 | 187 | 83 | ||
| BC138876.1 | - | 2 - 3 | 214 | 8861898 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox3
Genomic Map