Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 10 - 11 | 1034 | 126 | |
| GenBank mRNA | AF172276.2 | - | 10 - 11 | 1110 | 126 | |
| AK030109.1 | - | 6 - 7 | 574 | 126 | ||
| AK149434.1 | - | 10 - 11 | 995 | 126 | ||
| AY347569.1 | - | 10 - 11 | 911 | 126 | ||
| AY347570.1 | - | 10 - 11 | 911 | 126 | ||
| BC049853.1 | - | 10 - 11 | 986 | 126 | ||
| BC138876.1 | - | 10 - 11 | 1013 | 8861898 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox3
Genomic Map