Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 27 - 28 | 3281 | 102 | |
| GenBank mRNA | AF172276.2 | - | 27 - 28 | 3357 | 102 | |
| AK004700.1 | - | 16 - 17 | 2077 | 102 | ||
| AK030109.1 | - | 23 - 24 | 2821 | 102 | ||
| AK149434.1 | - | 27 - 28 | 3243 | 102 | ||
| AY347569.1 | - | 27 - 28 | 3158 | 102 | ||
| AY347570.1 | - | 27 - 28 | 3158 | 102 | ||
| BC049853.1 | - | 27 - 28 | 3233 | 102 | ||
| BC138876.1 | - | 27 - 28 | 3260 | 8861898 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox3
Genomic Map