Assay Details
| Gene Symbol: Aox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023631.2 | NP_076120.2 | 33 - 34 | 3969 | 73 | |
| GenBank mRNA | AF233581.1 | - | 33 - 34 | 3896 | 73 | |
| AK009112.1 | - | 8 - 9 | 888 | 73 | ||
| AK158427.1 | - | 33 - 34 | 3970 | 73 | ||
| BC117975.1 | - | 33 - 34 | 3896 | 40098789 | 73 | |
| BC117976.1 | - | 33 - 34 | 3896 | 40098791 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox4
Genomic Map