Assay Details
| Gene Symbol: Hdhd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024257.1 | NP_077219.1 | 1 - 2 | 40 | 73 | |
| GenBank mRNA | AK013245.1 | - | 1 - 2 | 55 | 73 | |
| BC003491.1 | - | 1 - 2 | 40 | 2811559 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK013245.1,BC003491.1,NM_024257.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964784 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964786 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964787 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964785 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P465582 | BC003491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P465582 | AK013245.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P465582 | NM_024257.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P465582 | BC003491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P465582 | AK013245.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P465582 | NM_024257.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 791756 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140137 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P465582 | BC003491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P465582 | AK013245.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P465582 | NM_024257.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P465582 | BC003491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P465582 | AK013245.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P465582 | NM_024257.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22964775 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140137 | - | - |
| Whole Mouse Genome G4122A | A_51_P465582 | BC003491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P465582 | AK013245.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P465582 | NM_024257.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P465582 | BC003491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P465582 | AK013245.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P465582 | NM_024257.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102626 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140137 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03516049 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03640996 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03516051 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964784 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964787 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964785 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22964786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22964806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04052032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03459142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03625930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03516049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03459143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03625937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03640997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03625924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03858163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03830413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03625944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03230658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03625913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03625914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03231746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03625927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03516054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03640996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22964796 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10513606 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513607 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425343_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10513604 | - | - |
| Mouse Genome 430 2.0 Array | 1425343_at | - | - |
| Mouse Genome 430 2.0 Array | 1425343_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72748_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72748_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400022904.mm.1 | TC0400002790.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400022904.mm.1 | NM_024257.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400022904.mm.1 | NM_024257.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400022906.mm.1 | TC0400002790.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400022906.mm.1 | NM_024257.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400022906.mm.1 | NM_024257.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400022905.mm.1 | TC0400002790.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400022905.mm.1 | NM_024257.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400022905.mm.1 | NM_024257.1 | Refseq |
| Mouse Expression Set 430 | 1425343_PM_at | - | - |
| Mouse Expression Set 430 | 1425343_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425343_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425343_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425343_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425343_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 745122 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6660132 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660132 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2608145 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2608145 | - | - |
| mouse-6 v1.1 (Illumina) | scl24262.3_152-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660132 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24262.3_152-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660132 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2608145 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2608145 | - | - |

Hdhd3
Genomic Map