Assay Details
Gene Symbol: Acy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276442.1 | NP_001263371.1 | 13 - 14 | 1115 | 76 | |
NM_025371.3 | NP_079647.1 | 13 - 14 | 1119 | 76 | ||
GenBank mRNA | AK002423.1 | - | 13 - 14 | 1105 | 76 | |
AK003703.1 | - | 13 - 14 | 1120 | 76 | ||
BC005631.1 | - | 13 - 14 | 1123 | 3964138 | 76 | |
BX529990.1 | - | 1 - 2 | 83 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P166288 | BC005631.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P166288 | NM_025371.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P166288 | AK002423.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P166288 | AK003703.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P166288 | BC005631.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P166288 | NM_025371.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P166288 | AK002423.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P166288 | AK003703.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953325 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250499 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547884 | AK021190.1 | genbank_mrna_mouse |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136797 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P166288 | BC005631.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P166288 | NM_025371.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P166288 | AK002423.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P166288 | AK003703.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P166288 | BC005631.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P166288 | NM_025371.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P166288 | AK002423.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P166288 | AK003703.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P166288 | BC005631.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P166288 | NM_025371.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P166288 | AK002423.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P166288 | AK003703.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P166288 | BC005631.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P166288 | NM_025371.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P166288 | AK002423.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P166288 | AK003703.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803372 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803377 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803367 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803347 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803359 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803364 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803344 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803352 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803348 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803354 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803366 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803356 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803374 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803362 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803342 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803360 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803350 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26803369 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02022888 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136797 | - | - |
Whole Mouse Genome G4122A | A_51_P166288 | BC005631.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P166288 | NM_025371.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P166288 | AK002423.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P166288 | AK003703.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P166288 | BC005631.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P166288 | NM_025371.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P166288 | AK002423.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P166288 | AK003703.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127269 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121092 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136797 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08005222 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08005228 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08005217 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08005219 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08005220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26803403 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803369 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08090144 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07664532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803316 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803364 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08005220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803331 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07664524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803321 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07664528 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803323 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07664533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07835094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803366 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07664530 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803359 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07952274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803360 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803344 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07574994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07574992 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803329 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803374 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803313 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08005219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803317 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803355 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803352 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07835097 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803338 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26803333 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10596478 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596468 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596476 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596470 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596473 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596474 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596477 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596475 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596469 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596472 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596471 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596479 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596466 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596467 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10596480 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419173_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10596465 | - | - |
Mouse Genome 430 2.0 Array | 1419173_at | - | - |
Mouse Genome 430 2.0 Array | 1419173_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 109652_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 109652_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900024974.mm.1 | NM_001276442.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024974.mm.1 | NM_025371.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900024974.mm.1 | TC0900003008.mm.1 | MTA TC |
Mouse Expression Set 430 | 1419173_PM_at | - | - |
Mouse Expression Set 430 | 1419173_at | - | - |
Murine Genome U74v2 Set | 101387_at | - | - |
Murine Genome U74v2 Set | 101387_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419173_at | - | - |
Mouse Genome 430A 2.0 Array | 1419173_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419173_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419173_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 601714 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2360279 | - | - |
MouseWG-6 v2 Expression BeadChip | 6550161 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6550161 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4480452 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2360279 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004480452 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006550161 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002360279 | - | - |
mouse-6 v1.1 (Illumina) | scl0003418.1_3-S | - | - |
mouse-6 v1.1 (Illumina) | scl35396.14.1_38-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4480452 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2360279 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6550161 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003418.1_3-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003579.1_12-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl35396.14.1_38-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360279 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480452 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550161 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2945472 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2757699 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213138 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2945472 | - | - |
