Assay Details
| Gene Symbol: Ccdc28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025455.2 | NP_079731.2 | 4 - 5 | 640 | 78 | |
| GenBank mRNA | AK007427.1 | - | 3 - 4 | 417 | 78 | |
| AK009176.1 | - | 2 - 3 | 260 | 78 | ||
| AK028262.1 | - | 4 - 5 | 618 | 78 | ||
| AK158016.1 | - | 4 - 5 | 644 | 78 | ||
| BC028873.1 | - | 4 - 5 | 632 | 4220355 | 78 | |
| BQ031123.1 | - | 2 - 3 | 78 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P102733 | AK045767.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P102733 | BC028873.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P102733 | BQ031123.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P102733 | NM_025455.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P102733 | AK009176.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429831 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130113 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156884 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P102733 | AK045767.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P102733 | BC028873.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P102733 | BQ031123.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P102733 | NM_025455.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P102733 | AK009176.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23297627 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156884 | - | - |
| Whole Mouse Genome G4122A | A_51_P495700 | NM_025455.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495700 | AK003287.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495700 | AK028262.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P112676 | BC028873.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P112676 | BQ031123.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P112676 | NM_025455.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P112676 | AK007427.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P112676 | AK009176.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120190 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102733 | AK045767.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102733 | BC028873.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102733 | BQ031123.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102733 | NM_025455.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102733 | AK009176.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156884 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03573495 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03248627 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03981841 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03573500 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03590663 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03590666 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03573506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23297537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03590663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03969260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03248627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03573507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03389937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03573503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03590665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03573512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03981835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03981844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03389931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03660573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23297649 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10516663 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516660 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516665 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516662 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516664 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516659 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516661 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452937_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459840_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442678_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430090_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10516658 | - | - |
| Mouse Genome 430 2.0 Array | 1442678_at | - | - |
| Mouse Genome 430 2.0 Array | 1430090_at | - | - |
| Mouse Genome 430 2.0 Array | 1430090_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442678_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452937_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1459840_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1459840_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1452937_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66264_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66264_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400029452.mm.1 | TC0400003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400029452.mm.1 | NM_025455.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029452.mm.1 | NM_025455.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029458.mm.1 | TC0400003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400029458.mm.1 | NM_025455.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029458.mm.1 | NM_025455.2 | Refseq |
| Mouse Expression Set 430 | 1452937_s_at | - | - |
| Mouse Expression Set 430 | 1459840_PM_s_at | - | - |
| Mouse Expression Set 430 | 1442678_at | - | - |
| Mouse Expression Set 430 | 1430090_at | - | - |
| Mouse Expression Set 430 | 1430090_PM_at | - | - |
| Mouse Expression Set 430 | 1442678_PM_at | - | - |
| Mouse Expression Set 430 | 1452937_PM_s_at | - | - |
| Mouse Expression Set 430 | 1459840_s_at | - | - |
| Murine Genome U74v2 Set | 160486_PM_at | - | - |
| Murine Genome U74v2 Set | 160486_at | - | - |
| Murine Genome U74v2 Set | 168461_at | - | - |
| Murine Genome U74v2 Set | 168461_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430090_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452937_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452937_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442678_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459840_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442678_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459840_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430090_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 930505 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5360433 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5360433 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4050563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005360433 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004050563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2694257 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220430 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2694257 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220430 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236759 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002652.1_12-S | - | - |
| mouse-6 v1.1 (Illumina) | scl066264.3_24-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5360433 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4050563 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066264.3_24-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002652.1_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4050563 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5360433 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1220430 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2694257 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220430 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2694257 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066264.3_24-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0002652.1_12-S | - | - |

Ccdc28b
Genomic Map