Assay Details
| Gene Symbol: Dhrs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025522.5 | NP_079798.2 | 4 - 5 | 694 | 72 | |
| GenBank mRNA | AK009385.1 | - | 4 - 5 | 673 | 72 | |
| AK014100.1 | - | 4 - 5 | 649 | 72 | ||
| BC016189.1 | - | 4 - 5 | 660 | 4482814 | 72 | |
| CF586466.1 | - | 3 - 4 | 72 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dhrs7
Genomic Map