Assay Details
Gene Symbol: Abi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025659.4 | NP_079935.1 | 1 - 2 | 420 | 76 | |
GenBank mRNA | AK008928.1 | - | 1 - 2 | 142 | 76 | |
AK018381.1 | - | 1 - 2 | 141 | 76 | ||
AK033965.1 | - | 1 - 2 | 141 | 76 | ||
AK172098.1 | - | 1 - 2 | 139 | 76 | ||
BC058260.1 | - | 1 - 2 | 130 | 6474610 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185906 | AK172098.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185906 | BC058260.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185906 | AK033965.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185906 | AK037144.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185906 | NM_001163464.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185906 | AK172098.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185906 | BC058260.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185906 | AK033965.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185906 | AK037144.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185906 | NM_001163464.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30854 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120032 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677420 | AK131750.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677420 | BC048573.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677420 | NM_001033362.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207145 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282188 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55476 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 778284 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171786 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143910 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185906 | AK172098.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185906 | BC058260.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185906 | AK033965.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185906 | AK037144.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185906 | NM_001163464.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185906 | AK172098.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185906 | BC058260.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185906 | AK033965.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185906 | AK037144.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185906 | NM_001163464.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185906 | AK172098.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185906 | BC058260.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185906 | AK033965.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185906 | AK037144.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185906 | NM_001163464.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185906 | AK172098.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185906 | BC058260.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185906 | AK033965.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185906 | AK037144.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P185906 | NM_001163464.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041140 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041132 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041114 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041135 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041129 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041138 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041137 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041130 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041133 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041123 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041128 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041126 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041119 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041139 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28041109 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02401644 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143910 | - | - |
Whole Mouse Genome G4122A | A_51_P185906 | AK172098.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P185906 | BC058260.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P185906 | AK033965.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P185906 | AK037144.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P185906 | NM_001163464.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P185906 | AK172098.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P185906 | BC058260.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P185906 | AK033965.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P185906 | AK037144.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P185906 | NM_001163464.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118633 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143910 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08982787 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08982779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28040994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28041125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041097 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09434283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09609062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08992217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041086 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041115 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041118 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09434281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041088 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09573579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041135 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09609066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09634371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041129 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09609060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09389785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041116 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28041096 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10390189 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390193 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390192 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390194 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390191 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390190 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390188 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390195 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1446191_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452928_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10380566 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10380571 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10390186 | - | - |
Mouse Genome 430 2.0 Array | 1446191_at | - | - |
Mouse Genome 430 2.0 Array | 1452928_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1452928_at | - | - |
Mouse Genome 430 2.0 Array | 1446191_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66610_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66610_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100032984.mm.1 | NM_001163464.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032984.mm.1 | NM_001163464.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032984.mm.1 | NM_025659.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032984.mm.1 | NM_025659.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032984.mm.1 | TC1100003600.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032988.mm.1 | NM_025659.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032988.mm.1 | NM_025659.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032988.mm.1 | TC1100003600.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032991.mm.1 | NM_025659.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032991.mm.1 | NM_025659.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032991.mm.1 | TC1100003600.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032989.mm.1 | NM_025659.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032989.mm.1 | NM_025659.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032989.mm.1 | TC1100003600.mm.1 | MTA TC |
Mouse Expression Set 430 | 1452928_at | - | - |
Mouse Expression Set 430 | 1446191_at | - | - |
Mouse Expression Set 430 | 1452928_PM_at | - | - |
Mouse Expression Set 430 | 1446191_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446191_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1452928_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1452928_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446191_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 931031 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1740563 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 101980162 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1740563 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001740563 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2616357 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2447169 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2616357 | - | - |
mouse-6 v1.1 (Illumina) | scl0066610.1_317-S | - | - |
mouse-6 v1.1 (Illumina) | scl39683.8_579-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 101980162 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1740563 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066610.1_317-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101980162 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740563 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1223041 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2447169 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223041 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0066610.1_317-S | - | - |
