Assay Details
Gene Symbol: Tmem170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025781.2 | NP_080057.1 | 2 - 3 | 349 | 63 | |
GenBank mRNA | AK007926.1 | - | 2 - 3 | 393 | 63 | |
AK018496.1 | - | 2 - 3 | 350 | 63 | ||
BC139377.1 | - | 2 - 3 | 424 | 8862399 | 63 | |
BC145964.1 | - | 2 - 3 | 424 | 40131267 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170718 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170721 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170720 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296429 | NM_025781.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296429 | AK018496.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296429 | NM_025781.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296429 | AK018496.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724674 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289935 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240685 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 733282 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 317873 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651088 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298794 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895787 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59272 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167003 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130982 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296429 | NM_025781.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296429 | AK018496.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296429 | NM_025781.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296429 | AK018496.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170722 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170714 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170706 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170720 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170731 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170712 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170711 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170718 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170704 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170728 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170723 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170721 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170708 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170715 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170717 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170716 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170703 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170726 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06643247 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167003 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130982 | - | - |
Whole Mouse Genome G4122A | A_51_P296429 | NM_025781.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296429 | AK018496.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296429 | NM_025781.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296429 | AK018496.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113869 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130982 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167003 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07167844 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07167836 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07167839 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170718 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170720 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07167839 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06932184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170717 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07167840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07195624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06930421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07189052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06821880 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170732 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170683 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170733 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06973770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170694 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170724 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07167836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06717747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07167835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06842003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06769706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07148586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06710467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170722 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10581911 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581912 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581913 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431128_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10581910 | - | - |
Mouse Genome 430 2.0 Array | 1439964_at | - | - |
Mouse Genome 430 2.0 Array | 1439964_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431128_at | - | - |
Mouse Genome 430 2.0 Array | 1431128_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66817_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66817_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800022841.mm.1 | NM_025781.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800022841.mm.1 | NM_025781.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800022841.mm.1 | TC0800002957.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800022840.mm.1 | NM_025781.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800022840.mm.1 | NM_025781.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800022840.mm.1 | TC0800002957.mm.1 | MTA TC |
Mouse Expression Set 430 | 1431128_at | - | - |
Mouse Expression Set 430 | 1431128_PM_at | - | - |
Murine Genome U74v2 Set | 139560_PM_at | - | - |
Murine Genome U74v2 Set | 139560_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431128_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431128_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 533225 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4590047 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004590047 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230101 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1230101 | - | - |
mouse-6 v1.1 (Illumina) | scl066817.1_216-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4590047 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl066817.1_216-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4590047 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2978655 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2978655 | - | - |
