Assay Details
| Gene Symbol: Use1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145780.1 | NP_001139252.1 | 8 - 9 | 861 | 62 | |
| NM_025917.4 | NP_080193.1 | 7 - 8 | 648 | 62 | ||
| GenBank mRNA | AF353245.1 | - | 7 - 8 | 641 | 62 | |
| AK003257.1 | - | 7 - 8 | 641 | 62 | ||
| AK003385.1 | - | 6 - 7 | 512 | 62 | ||
| AK008573.1 | - | 7 - 8 | 641 | 62 | ||
| AK008661.1 | - | 6 - 7 | 546 | 62 | ||
| AK012490.1 | - | 7 - 8 | 642 | 62 | ||
| AK017484.1 | - | 4 - 5 | 715 | 62 | ||
| AK160726.1 | - | 7 - 8 | 641 | 62 | ||
| AK165639.1 | - | 7 - 8 | 790 | 62 | ||
| AY842453.1 | - | 7 - 8 | 627 | 62 | ||
| BC025086.1 | - | 6 - 7 | 724 | 3995001 | 62 | |
| BC038519.1 | - | 6 - 7 | 711 | 1428854 | 62 | |
| BC075695.1 | - | 8 - 9 | 767 | 30623689 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956519 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956520 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956517 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967983 | AK165639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967983 | BC075695.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967983 | AK003257.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967983 | AK003385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967983 | AK008573.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810702 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298273 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147763 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121488 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178154 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171887 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136179 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967983 | AK165639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967983 | BC075695.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967983 | AK003257.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967983 | AK003385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967983 | AK008573.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P181235 | AK165639.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P181235 | AY842453.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P181235 | BC025086.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P181235 | BC038519.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P181235 | BC075695.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317076 | AK165639.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317076 | AY842453.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317076 | BC075695.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317076 | AK003257.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317076 | AK003385.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956513 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136179 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121488 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171887 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178154 | - | - |
| Whole Mouse Genome G4122A | A_51_P317076 | AK165639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317076 | AY842453.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317076 | BC075695.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317076 | AK003257.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317076 | AK003385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P181235 | AK165639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P181235 | AY842453.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P181235 | BC025086.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P181235 | BC038519.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P181235 | BC075695.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139950 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121488 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136179 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171887 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178154 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07137870 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07137852 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07273913 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07137856 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07137851 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07137869 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956517 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956520 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25956516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25956505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25956518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25956506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06750072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07273913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06777159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25956514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25956504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25956509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25956519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07312483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25956512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06794988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07137870 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10572586 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572587 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572584 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572583 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572590 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572589 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572585 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572581 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572582 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451130_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423817_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453430_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10572580 | - | - |
| Mouse Genome 430 2.0 Array | 1451130_at | - | - |
| Mouse Genome 430 2.0 Array | 1451130_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453430_at | - | - |
| Mouse Genome 430 2.0 Array | 1423817_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1453430_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423817_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67023_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67023_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800005619.mm.1 | NM_001145780.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800005619.mm.1 | NM_001145780.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800005619.mm.1 | NM_025917.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800005619.mm.1 | NM_025917.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800005619.mm.1 | TC0800000783.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800005620.mm.1 | NM_001145780.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800005620.mm.1 | NM_001145780.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800005620.mm.1 | NM_025917.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800005620.mm.1 | NM_025917.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800005620.mm.1 | TC0800000783.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451130_at | - | - |
| Mouse Expression Set 430 | 1423817_s_at | - | - |
| Mouse Expression Set 430 | 1423817_PM_s_at | - | - |
| Mouse Expression Set 430 | 1451130_PM_at | - | - |
| Murine Genome U74v2 Set | 99187_f_at | - | - |
| Murine Genome U74v2 Set | 99188_PM_at | - | - |
| Murine Genome U74v2 Set | 99188_at | - | - |
| Murine Genome U74v2 Set | 99187_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423817_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423817_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451130_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451130_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423817_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423817_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451130_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451130_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453430_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453430_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 915098 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480717 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1990338 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480717 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480717 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001990338 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2668778 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2667101 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2624385 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2624385 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2667101 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2668778 | - | - |
| mouse-6 v1.1 (Illumina) | scl000645.1_1-S | - | - |
| mouse-6 v1.1 (Illumina) | scl33705.5.1_110-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1990338 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480717 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000645.1_1-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33705.5.1_110-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480717 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990338 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2667101 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2624385 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2624385 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2667101 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33705.5.1_110-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000645.1_1-S | - | - |

Use1
Genomic Map