Assay Details
| Gene Symbol: Polr3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164082.1 | NP_001157554.1 | 2 - 3 | 325 | 71 | |
| NM_025945.3 | NP_080221.3 | 3 - 4 | 357 | 71 | ||
| GenBank mRNA | AK013165.1 | - | 2 - 3 | 242 | 71 | |
| AK156587.1 | - | 3 - 4 | 309 | 71 | ||
| BC016102.1 | - | 3 - 4 | 333 | 4911475 | 71 | |
| CA548436.1 | - | 2 - 3 | 274 | 71 | ||
| CD553533.1 | - | 3 - 4 | 339 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
transcription, DNA-templated
transcription from RNA polymerase III promoter
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity
DNA binding
chromatin binding
DNA-directed RNA polymerase activity
immune system process
transcription, DNA-templated
transcription from RNA polymerase III promoter
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity
DNA binding
chromatin binding
DNA-directed RNA polymerase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701298 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701301 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701291 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701299 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701295 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701300 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P223458 | AK156587.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P223458 | AW491400.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P223458 | BC016102.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P223458 | NM_025945.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P223458 | AK013165.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P223458 | AK156587.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P223458 | AW491400.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P223458 | BC016102.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P223458 | NM_025945.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P223458 | AK013165.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 622657 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175993 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149444 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P223458 | AK156587.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P223458 | AW491400.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P223458 | BC016102.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P223458 | NM_025945.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P223458 | AK013165.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P223458 | AK156587.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P223458 | AW491400.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P223458 | BC016102.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P223458 | NM_025945.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P223458 | AK013165.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P223458 | AK156587.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P223458 | AW491400.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P223458 | BC016102.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P223458 | NM_025945.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P223458 | AK013165.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P223458 | AK156587.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P223458 | AW491400.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P223458 | BC016102.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P223458 | NM_025945.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P223458 | AK013165.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701276 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149444 | - | - |
| Whole Mouse Genome G4122A | A_51_P223458 | AK156587.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223458 | AW491400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223458 | BC016102.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223458 | NM_025945.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223458 | AK013165.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223458 | AK156587.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223458 | AW491400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223458 | BC016102.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223458 | NM_025945.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223458 | AK013165.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114816 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149444 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11820851 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11820861 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11820856 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11820848 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11751723 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11820858 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11820849 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701291 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701300 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701295 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701301 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701299 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11421047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11820854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11379240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11692022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11421045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11820858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11291219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11820852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11820865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11751722 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10421358 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421360 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421355 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421356 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421359 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421357 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421354 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424565_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424566_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10421351 | - | - |
| Mouse Genome 430 2.0 Array | 1424566_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1424566_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1424565_at | - | - |
| Mouse Genome 430 2.0 Array | 1424565_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67065_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67065_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400018272.mm.1 | NM_001164082.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400018272.mm.1 | NM_001164082.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400018272.mm.1 | NM_025945.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400018272.mm.1 | NM_025945.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400018272.mm.1 | TC1400002341.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424566_s_at | - | - |
| Mouse Expression Set 430 | 1424566_PM_s_at | - | - |
| Mouse Expression Set 430 | 1424565_PM_at | - | - |
| Mouse Expression Set 430 | 1424565_at | - | - |
| Murine Genome U74v2 Set | 160650_at | - | - |
| Murine Genome U74v2 Set | 160650_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424565_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424566_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424565_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424566_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424565_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424566_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424565_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424566_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 819597 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6180364 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 580037 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006180364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000580037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230721 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215692 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215692 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230721 | - | - |
| mouse-6 v1.1 (Illumina) | scl067065.1_152-S | - | - |
| mouse-6 v1.1 (Illumina) | scl45352.5_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6180364 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 580037 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067065.1_152-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl45352.5_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580037 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6180364 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1215692 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1230721 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230721 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215692 | - | - |

Polr3d
Genomic Map