Assay Details
| Gene Symbol: Dynlrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291108.1 | NP_001278037.1 | 2 - 3 | 407 | 59 | |
| NM_025947.3 | NP_080223.2 | 2 - 3 | 163 | 59 | ||
| NR_111890.1 | 2 - 3 | 411 | 59 | |||
| NR_111891.1 | 2 - 3 | 159 | 59 | |||
| GenBank mRNA | AK002359.1 | - | 2 - 3 | 104 | 59 | |
| AK003435.1 | - | 1 - 2 | 93 | 59 | ||
| AK008595.1 | - | 3 - 4 | 125 | 59 | ||
| AK166685.1 | - | 2 - 3 | 139 | 59 | ||
| BC099423.1 | - | 2 - 3 | 123 | 30933448 | 59 | |
| BC125610.1 | - | 2 - 3 | 123 | 40130125 | 59 | |
| BC125648.1 | - | 2 - 3 | 123 | 40130163 | 59 | |
| CJ136086.1 | - | 2 - 3 | 408 | 59 | ||
| CO041603.1 | - | 2 - 3 | 59 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764579 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764583 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764585 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764581 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764584 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069949 | AK008595.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069949 | AK166685.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069949 | BC099423.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069949 | CO041603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069949 | NM_025947.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105981 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569498 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147155 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56827 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136242 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151530 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069949 | AK008595.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069949 | AK166685.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069949 | BC099423.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069949 | CO041603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069949 | NM_025947.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P254553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21764588 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04748916 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151530 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136242 | - | - |
| Whole Mouse Genome G4122A | A_51_P254553 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121251 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151530 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136242 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01392822 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01173264 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01173268 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764584 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764581 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764579 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764583 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21764585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21764656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01944374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01918965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01918957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02271132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01392822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01557462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01531863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02162491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01918960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02093639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02162489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01459377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01380431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01944375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01173275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01757315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01918964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01380433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02271138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02162490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01385216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01558668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01898948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01531865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21764574 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10477634 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477631 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477635 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428257_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10477630 | - | - |
| Mouse Genome 430 2.0 Array | 1428257_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1428257_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67068_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67068_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200018811.mm.1 | NM_025947.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018811.mm.1 | NM_025947.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018811.mm.1 | TC0200002362.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200018813.mm.1 | NM_025947.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018813.mm.1 | NM_025947.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018813.mm.1 | TC0200002362.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200018820.mm.1 | NM_025947.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018820.mm.1 | NM_025947.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018820.mm.1 | TC0200002362.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200018819.mm.1 | NM_025947.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018819.mm.1 | NM_025947.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018819.mm.1 | TC0200002362.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428257_PM_s_at | - | - |
| Mouse Expression Set 430 | 1428257_s_at | - | - |
| Murine Genome U74v2 Set | 94862_PM_i_at | - | - |
| Murine Genome U74v2 Set | 94863_PM_r_at | - | - |
| Murine Genome U74v2 Set | 94863_r_at | - | - |
| Murine Genome U74v2 Set | 94862_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428257_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428257_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428257_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428257_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 928949 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6760358 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6760358 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102640139 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006760358 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2589814 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2491303 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2589814 | - | - |
| mouse-6 v1.1 (Illumina) | scl077350.1_78-S | - | - |
| mouse-6 v1.1 (Illumina) | scl20055.4.1_10-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102640139 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6760358 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20055.4.1_10-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102640139 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6760358 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2589814 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2491303 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2589814 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl20055.4.1_10-S | - | - |

Dynlrb1
Genomic Map