Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500024266.mm.1 |
TC0500002753.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024266.mm.1 |
NM_001163687.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024266.mm.1 |
NM_001163687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024266.mm.1 |
NM_025972.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024266.mm.1 |
NM_025972.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024268.mm.1 |
TC0500002753.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024268.mm.1 |
NM_001163687.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024268.mm.1 |
NM_001163687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024268.mm.1 |
NM_025972.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024268.mm.1 |
NM_025972.4 |
Refseq |