Assay Details
| Gene Symbol: Glod4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026029.3 | NP_080305.2 | 4 - 5 | 456 | 86 | |
| GenBank mRNA | AK006979.1 | - | 4 - 5 | 779 | 86 | |
| AK007439.1 | - | 4 - 5 | 421 | 86 | ||
| AK010992.1 | - | 4 - 5 | 414 | 86 | ||
| AK012569.1 | - | 4 - 5 | 434 | 86 | ||
| AK013212.1 | - | 4 - 5 | 434 | 86 | ||
| AK138073.1 | - | 433 | 86 | |||
| BC049937.1 | - | 1 - 2 | 85 | 86 | ||
| BC061012.1 | - | 4 - 5 | 448 | 6587019 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glod4
Genomic Map