Assay Details
| Gene Symbol: Glod4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026029.3 | NP_080305.2 | 5 - 6 | 597 | 81 | |
| GenBank mRNA | AK006979.1 | - | 5 - 6 | 920 | 81 | |
| AK007439.1 | - | 5 - 6 | 562 | 81 | ||
| AK010992.1 | - | 5 - 6 | 555 | 81 | ||
| AK012569.1 | - | 5 - 6 | 575 | 81 | ||
| AK013212.1 | - | 5 - 6 | 575 | 81 | ||
| AK138073.1 | - | 574 | 81 | |||
| BC049937.1 | - | 2 - 3 | 226 | 81 | ||
| BC061012.1 | - | 5 - 6 | 589 | 6587019 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glod4
Genomic Map