Assay Details
| Gene Symbol: Glod4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026029.3 | NP_080305.2 | 7 - 8 | 795 | 68 | |
| GenBank mRNA | AK006979.1 | - | 7 - 8 | 1118 | 68 | |
| AK007439.1 | - | 7 - 8 | 760 | 68 | ||
| AK010992.1 | - | 7 - 8 | 753 | 68 | ||
| AK012569.1 | - | 7 - 8 | 773 | 68 | ||
| AK013212.1 | - | 7 - 8 | 773 | 68 | ||
| AK033042.1 | - | 3 - 4 | 226 | 68 | ||
| AK138073.1 | - | 772 | 68 | |||
| BC049937.1 | - | 4 - 5 | 424 | 68 | ||
| BC055356.1 | - | 2 - 3 | 168 | 68 | ||
| BC061012.1 | - | 7 - 8 | 787 | 6587019 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glod4
Genomic Map