Assay Details
| Gene Symbol: Mboat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083341.1 | NP_001076810.1 | 7 - 8 | 885 | 68 | |
| NM_026037.3 | NP_080313.2 | 8 - 9 | 981 | 68 | ||
| GenBank mRNA | AB297383.1 | - | 8 - 9 | 880 | 68 | |
| AB305046.1 | - | 8 - 9 | 880 | 68 | ||
| AK012931.1 | - | 7 - 8 | 869 | 68 | ||
| AK031824.1 | - | 7 - 8 | 870 | 68 | ||
| AK033106.1 | - | 5 - 6 | 645 | 68 | ||
| AK034873.1 | - | 5 - 6 | 669 | 68 | ||
| AK076045.1 | - | 7 - 8 | 875 | 68 | ||
| BC025020.1 | - | 7 - 8 | 835 | 5364068 | 68 | |
More Information
Set Membership: |
|

Mboat2
Genomic Map