Assay Details
Gene Symbol: Gemin6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026053.3 | NP_080329.1 | 1 - 2 | 188 | 70 | |
GenBank mRNA | AK011454.1 | - | 1 - 2 | 169 | 70 | |
AK013396.1 | - | 1 - 2 | 190 | 70 | ||
AK018385.1 | - | 1 - 2 | 185 | 70 | ||
AK020147.1 | - | 1 - 2 | 212 | 70 | ||
AK157745.1 | - | 1 - 2 | 201 | 70 | ||
AK167920.1 | - | 1 - 2 | 227 | 70 | ||
BC025157.1 | - | 1 - 2 | 171 | 3670793 | 70 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK011454.1,AK013396.1,AK018385.1,AK020147.1,AK157745.1,AK167920.1,BC025157.1,NM_026053.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095784 | AK013396.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095784 | NM_026053.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093727 | AK167920.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093727 | BC025157.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093727 | AK011454.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093727 | AK020147.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093727 | NM_026053.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494247 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595115 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 58522 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167413 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095784 | AK013396.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095784 | NM_026053.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093727 | AK167920.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093727 | BC025157.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093727 | AK011454.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093727 | AK020147.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093727 | NM_026053.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P190745 | AK167920.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P190745 | BC025157.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P190745 | AK011454.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P190745 | AK013396.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P190745 | AK018385.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381829 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381826 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381844 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381836 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381854 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381830 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381843 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381848 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381827 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381856 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381823 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381819 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381833 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381818 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381821 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381846 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381840 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381838 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31381825 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167413 | - | - |
Whole Mouse Genome G4122A | A_52_P190745 | AK167920.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P190745 | BC025157.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P190745 | AK011454.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P190745 | AK013396.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P190745 | AK018385.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126989 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167413 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13342857 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13139211 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13139215 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13586742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31381885 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381876 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13292418 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381817 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13545189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381846 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381796 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381809 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381857 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381838 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381880 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13586742 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381829 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381843 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13292437 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381865 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13292422 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381854 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13498154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381832 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13105698 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13521332 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381812 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381804 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381872 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13292416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381806 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381856 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381823 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381819 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381807 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381844 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13498155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13498733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381827 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381879 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13139216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381799 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381813 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381830 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381877 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13366856 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13332719 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381831 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381818 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381878 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31381849 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10541092 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10447099 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541093 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10447098 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424300_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10447097 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10541091 | - | - |
Mouse Genome 430 2.0 Array | 1424300_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1424300_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67242_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67242_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700011362.mm.1 | NM_026053.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700011362.mm.1 | TC1700001211.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700011364.mm.1 | NM_026053.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700011364.mm.1 | TC1700001211.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700011360.mm.1 | NM_026053.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700011360.mm.1 | TC1700001211.mm.1 | MTA TC |
Mouse Expression Set 430 | 1424300_PM_at | - | - |
Mouse Expression Set 430 | 1424300_at | - | - |
Mouse Genome 430A 2.0 Array | 1424300_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1424300_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424300_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424300_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 898923 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2120288 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3830176 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003830176 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002120288 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214319 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2641340 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2641340 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1214319 | - | - |
mouse-6 v1.1 (Illumina) | scl0067242.1_20-S | - | - |
mouse-6 v1.1 (Illumina) | scl067242.3_45-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2120288 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3830176 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl067242.3_45-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067242.1_20-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2120288 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3830176 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2631813 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1214319 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2641340 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2641340 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631813 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214319 | - | - |
