Assay Details
| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026194.4 | NP_080470.2 | 4 - 5 | 415 | 103 | |
| GenBank mRNA | AK007993.2 | - | 4 - 5 | 360 | 103 | |
| AK032663.1 | - | 4 - 5 | 351 | 103 | ||
| AK076206.1 | - | 4 - 5 | 360 | 103 | ||
| AK129211.1 | - | 3 - 4 | 328 | 103 | ||
| AK132697.1 | - | 4 - 5 | 379 | 103 | ||
| BC058809.1 | - | 4 - 5 | 354 | 103 | ||
| BC080203.1 | - | 4 - 5 | 393 | 103 | ||
| BC099529.1 | - | 4 - 5 | 403 | 103 | ||
| BC132195.1 | - | 4 - 5 | 367 | 40130472 | 103 | |
| BC138153.1 | - | 4 - 5 | 367 | 8861172 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ufl1
Genomic Map