Assay Details
Gene Symbol: Tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026210.4 | NP_080486.1 | 5 - 6 | 557 | 57 | |
GenBank mRNA | AK007707.1 | - | 5 - 6 | 473 | 57 | |
AK017572.1 | - | 5 - 6 | 472 | 57 | ||
AK050027.1 | - | 5 - 6 | 593 | 57 | ||
AK077277.1 | - | 5 - 6 | 559 | 57 | ||
AK150936.1 | - | 5 - 6 | 549 | 57 | ||
BC115504.1 | - | 5 - 6 | 537 | 40101013 | 57 | |
BC115505.1 | - | 5 - 6 | 537 | 40101015 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856223 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856224 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856227 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856228 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856225 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856226 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046699 | AK150936.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046699 | AK007707.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046699 | AK017572.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046699 | AK077277.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046699 | NM_026210.4 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459848 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437558 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563985 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671636 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 733504 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385415 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676169 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809535 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646973 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634135 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13666 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175767 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149658 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046699 | AK150936.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046699 | AK007707.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046699 | AK017572.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046699 | AK077277.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046699 | NM_026210.4 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856220 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856235 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856217 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856225 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856213 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856231 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856216 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856222 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856208 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856241 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856239 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856232 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856210 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856218 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856234 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856207 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856238 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856227 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856230 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27856240 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149658 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175767 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07368819 | - | - |
Whole Mouse Genome G4122A | A_51_P287206 | AK150936.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P287206 | AK007707.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P287206 | AK017572.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P287206 | AK077277.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P287206 | NM_026210.4 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106045 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109446 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175767 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149658 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09088804 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09276455 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09088810 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09088816 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09088819 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856223 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856225 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856224 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856227 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856228 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27856226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27856331 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856241 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09148034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09423267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09113435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09625466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09088813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09088807 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09113433 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09605597 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856261 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856254 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856218 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856271 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856273 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09276434 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856277 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09406202 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856260 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856210 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09276456 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09088812 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856259 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09625468 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09625474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09088817 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27856208 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10376932 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10376930 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10376931 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10376937 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10376933 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10376935 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10376936 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10376934 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452953_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10376929 | - | - |
Mouse Genome 430 2.0 Array | 1452953_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1452953_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67510_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67510_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100006578.mm.1 | NM_026210.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100006578.mm.1 | NM_026210.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100006578.mm.1 | TC1100000816.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100006581.mm.1 | NM_026210.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100006581.mm.1 | NM_026210.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100006581.mm.1 | TC1100000816.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100006582.mm.1 | NM_026210.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100006582.mm.1 | NM_026210.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100006582.mm.1 | TC1100000816.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100006579.mm.1 | NM_026210.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100006579.mm.1 | NM_026210.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100006579.mm.1 | TC1100000816.mm.1 | MTA TC |
Mouse Expression Set 430 | 1452953_at | - | - |
Mouse Expression Set 430 | 1452953_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1452953_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1452953_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 430905 | - | - |
ABI Mouse Genome Survey Microarray | 758511 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2120671 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2120671 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002120671 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2665946 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2665946 | - | - |
mouse-6 v1.1 (Illumina) | scl41439.7_320-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2120671 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl41439.7_320-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2120671 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2665943 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2665943 | - | - |
