Assay Details
Gene Symbol: Patl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026251.2 | NP_080527.2 | 12 - 13 | 1408 | 77 | |
GenBank mRNA | AK015195.1 | - | 12 - 13 | 1412 | 77 | |
BC145646.1 | - | 10 - 11 | 1189 | 40130949 | 77 | |
DV660077.1 | - | 3 - 4 | 272 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
