Assay Details
Gene Symbol: Patl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026251.2 | NP_080527.2 | 13 - 14 | 1547 | 109 | |
GenBank mRNA | AK015195.1 | - | 13 - 14 | 1551 | 109 | |
BC145646.1 | - | 11 - 12 | 1328 | 40130949 | 109 | |
DV660077.1 | - | 4 - 5 | 411 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
