Assay Details
| Gene Symbol: Iah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026347.3 | NP_080623.2 | 1 - 2 | 117 | 70 | |
| GenBank mRNA | AK005287.1 | - | 1 - 2 | 118 | 70 | |
| AK011299.1 | - | 1 - 2 | 118 | 70 | ||
| AK014743.1 | - | 1 - 2 | 95 | 70 | ||
| BC060949.1 | - | 1 - 2 | 120 | 6533986 | 70 | |
| BC087901.1 | - | 1 - 2 | 88 | 30303437 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
biological_process
lipid catabolic process
molecular_function
nuclease activity
hydrolase activity
hydrolase activity, acting on ester bonds
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
lipid metabolic process
biological_process
lipid catabolic process
molecular_function
nuclease activity
hydrolase activity
hydrolase activity, acting on ester bonds
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28280007 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28280008 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28280005 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507305 | AK014743.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507305 | AK005287.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507305 | AK011299.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507305 | BC060949.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507305 | BC087901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507305 | AK014743.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507305 | AK005287.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507305 | AK011299.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507305 | BC060949.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P507305 | BC087901.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 598226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 26503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84207 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295611 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507305 | AK014743.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507305 | AK005287.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507305 | AK011299.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507305 | BC060949.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507305 | BC087901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507305 | AK014743.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507305 | AK005287.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507305 | AK011299.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507305 | BC060949.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P507305 | BC087901.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28280010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279998 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07556790 | - | - |
| Whole Mouse Genome G4122A | A_51_P281637 | AK014743.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P507305 | AK014743.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P507305 | AK005287.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P507305 | AK011299.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P507305 | BC060949.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P507305 | BC087901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P507305 | AK014743.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P507305 | AK005287.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P507305 | AK011299.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P507305 | BC060949.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P507305 | BC087901.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109061 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130353 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10282980 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10282993 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10282989 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10091841 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10282975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28280007 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28280005 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28280008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28280001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09933379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10282980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10282112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10282111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10282976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10237084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10282978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10237083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10091841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09821510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10282990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09823743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10282972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09962535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10282996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09821516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28280002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10402216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10282975 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10394915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10394919 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429119_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454898_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453471_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10394914 | - | - |
| Mouse Genome 430 2.0 Array | 1429119_at | - | - |
| Mouse Genome 430 2.0 Array | 1454898_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429119_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453471_at | - | - |
| Mouse Genome 430 2.0 Array | 1454898_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1453471_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67732_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67732_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200001464.mm.1 | NM_026347.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001464.mm.1 | NM_026347.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001464.mm.1 | TC1200000183.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200001465.mm.1 | NM_026347.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001465.mm.1 | NM_026347.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001465.mm.1 | TC1200000183.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200001466.mm.1 | NM_026347.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001466.mm.1 | NM_026347.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200001466.mm.1 | TC1200000183.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454898_PM_s_at | - | - |
| Mouse Expression Set 430 | 1454898_s_at | - | - |
| Mouse Expression Set 430 | 1453471_PM_at | - | - |
| Mouse Expression Set 430 | 1453471_at | - | - |
| Murine Genome U74v2 Set | 167919_at | - | - |
| Murine Genome U74v2 Set | 167919_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454898_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454898_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454898_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454898_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429119_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429119_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453471_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453471_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 915218 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2900170 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1690309 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2900170 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002900170 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001690309 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2723208 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2620803 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2620803 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2723208 | - | - |
| mouse-6 v1.1 (Illumina) | scl0067732.1_253-S | - | - |
| mouse-6 v1.1 (Illumina) | scl067732.2_80-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2900170 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1690309 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067732.1_253-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067732.2_80-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900170 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1690309 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2620803 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2620804 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2620803 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2620804 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0067732.1_253-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067732.2_80-S | - | - |

Iah1
Genomic Map