Assay Details
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026485.2 | NP_080761.2 | 4 - 5 | 389 | 78 | |
GenBank mRNA | AK005300.1 | - | 4 - 5 | 359 | 78 | |
AK017745.1 | - | 4 - 5 | 383 | 78 | ||
AK146027.1 | - | 4 - 5 | 405 | 78 | ||
BC005574.1 | - | 4 - 5 | 415 | 3967961 | 78 | |
BU510197.1 | - | 4 - 5 | 469 | 78 | ||
CX200611.1 | - | 4 - 5 | 374 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
