Assay Details
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026485.2 | NP_080761.2 | ||||
GenBank mRNA | AK005300.1 | - | ||||
AK017745.1 | - | |||||
AK146027.1 | - | |||||
BC005574.1 | - | 3967961 | ||||
BU510197.1 | - | |||||
CX200611.1 | - |

More Information
Updated Mapping Information

Set Membership: |
![]() |
Gene Ontology Categories:
MicroArray Probes:
