Assay Details
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026485.2 | NP_080761.2 | 6 - 7 | 642 | 68 | |
GenBank mRNA | AK005300.1 | - | 6 - 7 | 612 | 68 | |
AK017745.1 | - | 6 - 7 | 636 | 68 | ||
AK146027.1 | - | 6 - 7 | 658 | 68 | ||
BC005574.1 | - | 6 - 7 | 668 | 3967961 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
