Assay Details
| Gene Symbol: Toe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026654.2 | NP_080930.1 | 2 - 3 | 235 | 93 | |
| GenBank mRNA | AK016889.1 | - | 225 | 93 | ||
| AK019829.1 | - | 2 - 3 | 235 | 93 | ||
| AK046576.1 | - | 2 - 3 | 266 | 93 | ||
| AK082975.1 | - | 2 - 3 | 216 | 93 | ||
| BC023109.1 | - | 2 - 3 | 211 | 4021435 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Toe1
Genomic Map