Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300012026.mm.1 |
NM_026656.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012026.mm.1 |
NM_026656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012026.mm.1 |
TC0300001489.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012027.mm.1 |
NM_026656.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012027.mm.1 |
NM_026656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012027.mm.1 |
TC0300001489.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012030.mm.1 |
NM_001005846.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012030.mm.1 |
NM_001005846.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012030.mm.1 |
NM_026656.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012030.mm.1 |
NM_026656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012030.mm.1 |
TC0300001489.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012031.mm.1 |
NM_001005846.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012031.mm.1 |
NM_001005846.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012031.mm.1 |
NM_026656.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012031.mm.1 |
NM_026656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012031.mm.1 |
TC0300001489.mm.1 |
MTA TC |