Assay Details
| Gene Symbol: Med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290489.1 | NP_001277418.1 | 5 - 6 | 484 | 94 | |
| NM_026896.5 | NP_081172.2 | 6 - 7 | 758 | 94 | ||
| GenBank mRNA | AK005251.1 | - | 6 - 7 | 733 | 94 | |
| BC016537.1 | - | 6 - 7 | 749 | 3585027 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med27
Genomic Map