Assay Details
Gene Symbol: Rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026967.4 | NP_081243.1 | 4 - 5 | 488 | 90 | |
GenBank mRNA | AK007708.1 | - | 4 - 5 | 338 | 90 | |
AK135858.1 | - | 4 - 5 | 443 | 90 | ||
AY327413.1 | - | 4 - 5 | 536 | 90 | ||
BC016521.1 | - | 4 - 5 | 407 | 3487009 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
