Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400002582.mm.1 |
NM_028839.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002582.mm.1 |
NM_028839.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002582.mm.1 |
TC1400000314.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002585.mm.1 |
TC1400000314.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002588.mm.1 |
NM_028839.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002588.mm.1 |
NM_028839.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002588.mm.1 |
TC1400000314.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002583.mm.1 |
NM_028839.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002583.mm.1 |
NM_028839.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002583.mm.1 |
TC1400000314.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002584.mm.1 |
NM_028839.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002584.mm.1 |
NM_028839.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002584.mm.1 |
TC1400000314.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002586.mm.1 |
NM_028839.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002586.mm.1 |
NM_028839.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002586.mm.1 |
TC1400000314.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002589.mm.1 |
NM_028839.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002589.mm.1 |
NM_028839.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002589.mm.1 |
TC1400000314.mm.1 |
MTA TC |