Assay Details
Gene Symbol: Lrrc46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027026.2 | NP_081302.2 | 1 - 2 | 290 | 81 | |
GenBank mRNA | AK005668.1 | - | 1 - 2 | 266 | 81 | |
AK158517.1 | - | 1 - 2 | 337 | 81 | ||
BC022723.1 | - | 1 - 2 | 290 | 4503387 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK005668.1,AK158517.1,BC022723.1,NM_027026.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
