Assay Details
| Gene Symbol: Efcab9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027031.3 | NP_081307.2 | 1 - 2 | 191 | 95 | |
| GenBank mRNA | AK005716.1 | - | 1 - 2 | 196 | 95 | |
| AK015057.1 | - | 1 - 2 | 182 | 95 | ||
| AK161300.1 | - | 1 - 2 | 182 | 95 | ||
| BC087911.1 | - | 1 - 2 | 191 | 6771929 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27689365 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27689366 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27689364 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139029 | AK161300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139029 | BC087911.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139029 | NM_027031.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139029 | AK005716.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420355 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173397 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894760 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104969 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139029 | AK161300.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139029 | BC087911.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139029 | NM_027031.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139029 | AK005716.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27689334 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104969 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07296822 | - | - |
| Whole Mouse Genome G4122A | A_51_P251707 | AK161300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P251707 | BC087911.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P251707 | NM_027031.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P251707 | AK005716.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P251707 | AK015057.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139029 | AK161300.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139029 | BC087911.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139029 | NM_027031.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139029 | AK005716.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104969 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09395668 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09395670 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09282052 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09507688 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09395677 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09244241 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09507681 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09395667 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09507684 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27689366 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27689364 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27689365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27689386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09572324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09499995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09572326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09507681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09443949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09582939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09244243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09507682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09507684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09572341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27689318 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10385026 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385024 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385023 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385025 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429746_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10385022 | - | - |
| Mouse Genome 430 2.0 Array | 1429746_at | - | - |
| Mouse Genome 430 2.0 Array | 1429746_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69306_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69306_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100023088.mm.1 | NM_027031.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100023088.mm.1 | NM_027031.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100023088.mm.1 | TC1100002476.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100023089.mm.1 | NM_027031.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100023089.mm.1 | NM_027031.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100023089.mm.1 | TC1100002476.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429746_at | - | - |
| Mouse Expression Set 430 | 1429746_PM_at | - | - |
| Murine Genome U74v2 Set | 166287_PM_f_at | - | - |
| Murine Genome U74v2 Set | 166287_f_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429746_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429746_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 448648 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2320021 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002320021 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2647458 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2647458 | - | - |
| mouse-6 v1.1 (Illumina) | scl40388.5.1_14-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2320021 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40388.5.1_14-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320021 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2647458 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2647458 | - | - |

Efcab9
Genomic Map