Assay Details
Gene Symbol: Hmgb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027036.3 | NP_081312.2 | 1 | 636 | 74 | |
GenBank mRNA | AK005588.1 | - | 1 | 635 | 74 | |
BC048744.1 | - | 2 | 596 | 74 | ||
BC061030.1 | - | 1 | 632 | 6775221 | 74 | |
BC087910.1 | - | 2 | 522 | 74 | ||
BC087934.1 | - | 2 | 592 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380919 | CN840046.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380919 | NM_027036.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380919 | AK005588.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380919 | BC048744.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380919 | BC061030.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380919 | CN840046.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380919 | NM_027036.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380919 | AK005588.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380919 | BC048744.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P380919 | BC061030.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148736 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380919 | CN840046.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380919 | NM_027036.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380919 | AK005588.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380919 | BC048744.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380919 | BC061030.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380919 | CN840046.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380919 | NM_027036.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380919 | AK005588.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380919 | BC048744.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P380919 | BC061030.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289839 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289837 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289831 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289832 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289846 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289806 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289829 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289802 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289842 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289838 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289834 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289808 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289803 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289812 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289843 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289817 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289826 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289809 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289848 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289820 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289853 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289810 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289819 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289813 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289816 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289822 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289852 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289824 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23289825 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148736 | - | - |
Whole Mouse Genome G4122A | A_51_P380919 | CN840046.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P380919 | NM_027036.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P380919 | AK005588.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P380919 | BC048744.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P380919 | BC061030.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P380919 | CN840046.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P380919 | NM_027036.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P380919 | AK005588.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P380919 | BC048744.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P380919 | BC061030.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111970 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148736 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04084585 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04084576 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04084582 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04084592 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04084589 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04084586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23289777 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289874 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03414730 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289820 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03712758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289831 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04004630 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289796 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289791 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04004627 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289768 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04084594 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289822 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289794 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04061882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03438000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289851 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289872 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289835 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289817 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289760 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03984391 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289763 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289789 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289800 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04061866 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03286071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289824 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289780 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03248982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289766 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289826 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289853 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289787 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04057804 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04061860 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289786 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289816 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04061886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289840 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04004619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289850 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03437997 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289812 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03939549 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289792 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03637482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289863 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289809 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289833 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289782 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289803 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289797 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289869 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23289842 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03233064 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03286070 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10516505 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516503 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516504 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10516506 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429349_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10516502 | - | - |
Mouse Genome 430 2.0 Array | 1429349_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1429349_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69317_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69317_PM_at | - | - |
Mouse Expression Set 430 | 1429349_PM_at | - | - |
Mouse Expression Set 430 | 1429349_at | - | - |
Murine Genome U74v2 Set | 103632_at | - | - |
Murine Genome U74v2 Set | 103632_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1429349_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1429349_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1429349_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1429349_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 401226 | - | - |
ABI Mouse Genome Survey Microarray | 702135 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5130373 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5130373 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005130373 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2741149 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2741149 | - | - |
mouse-6 v1.1 (Illumina) | scl23798.1_30-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5130373 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl23798.1_30-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130373 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2741149 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2741149 | - | - |
