Assay Details
| Gene Symbol: Dennd6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027081.3 | NP_081357.2 | 17 - 18 | 1535 | 77 | |
| GenBank mRNA | AK006427.1 | - | 8 - 9 | 672 | 77 | |
| AK084555.1 | - | 15 - 16 | 1561 | 77 | ||
| BC017544.1 | - | 9 - 10 | 1491 | 77 | ||
| BC058958.1 | - | 13 - 14 | 1042 | 77 | ||
| BC080797.1 | - | 13 - 14 | 1424 | 77 | ||
| BC139003.1 | - | 17 - 18 | 1509 | 8862025 | 77 | |
| BQ176037.1 | - | 3 - 4 | 77 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd6b
Genomic Map