Assay Details
Gene Symbol: Dnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172154.1 | NP_001165625.1 | 6 - 7 | 778 | 76 | |
NM_027109.2 | NP_081385.1 | 6 - 7 | 895 | 76 | ||
GenBank mRNA | AK009174.1 | - | 6 - 7 | 901 | 76 | |
AK150242.1 | - | 6 - 7 | 808 | 76 | ||
BC023246.1 | - | 6 - 7 | 778 | 4039470 | 76 | |
DQ116784.1 | - | 6 - 7 | 789 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032128 | AK150242.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032128 | BC023246.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032128 | DQ116784.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032128 | NM_027109.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032128 | AK009174.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960311 | AK150242.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960311 | BC023246.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960311 | DQ116784.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960311 | NM_027109.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960311 | AK009174.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357127 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380450 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72221 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771496 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794642 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951507 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356118 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11309 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155429 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154450 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960311 | AK150242.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960311 | BC023246.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960311 | DQ116784.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960311 | NM_027109.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960311 | AK009174.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032128 | AK150242.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032128 | BC023246.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032128 | DQ116784.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032128 | NM_027109.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032128 | AK009174.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P236769 | AK150242.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P236769 | BC023246.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P236769 | DQ116784.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P236769 | NM_027109.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P236769 | AK009174.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467241 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467233 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467235 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467209 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467232 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467256 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467234 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467238 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09331255 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155429 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03698288 | - | - |
Whole Mouse Genome G4122A | A_51_P236769 | AK150242.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P236769 | BC023246.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P236769 | DQ116784.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P236769 | NM_027109.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P236769 | AK009174.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117797 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155429 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14720303 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14720274 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14720298 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14720296 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14720275 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14720270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467250 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467209 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467243 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14733432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14720303 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467226 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467212 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14593730 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14664484 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14720297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467238 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467249 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467222 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14664488 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14733415 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15129354 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467229 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15129353 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14720289 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467215 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14733409 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467241 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467239 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467253 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14720294 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10605308 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10605306 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10605309 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10605304 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10605313 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10605314 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10605311 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10605312 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10605307 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10605310 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429173_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10605303 | - | - |
Mouse Genome 430 2.0 Array | 1429173_at | - | - |
Mouse Genome 430 2.0 Array | 1429173_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69537_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69537_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00015548.mm.1 | NM_001172154.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015548.mm.1 | NM_001172154.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015548.mm.1 | NM_027109.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015548.mm.1 | NM_027109.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015548.mm.1 | TC0X00002448.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00015549.mm.1 | NM_001172154.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015549.mm.1 | NM_001172154.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015549.mm.1 | NM_027109.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015549.mm.1 | NM_027109.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015549.mm.1 | TC0X00002448.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00015551.mm.1 | NM_001172154.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015551.mm.1 | NM_001172154.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015551.mm.1 | NM_027109.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015551.mm.1 | NM_027109.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015551.mm.1 | TC0X00002448.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00015550.mm.1 | NM_001172154.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015550.mm.1 | NM_001172154.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015550.mm.1 | NM_027109.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015550.mm.1 | NM_027109.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00015550.mm.1 | TC0X00002448.mm.1 | MTA TC |
Mouse Expression Set 430 | 1429173_at | - | - |
Mouse Expression Set 430 | 1429173_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1429173_at | - | - |
Mouse Genome 430A 2.0 Array | 1429173_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1429173_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1429173_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 400553 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6420066 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6420066 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006420066 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213982 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1213982 | - | - |
mouse-6 v1.1 (Illumina) | scl54140.7.1_25-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6420066 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl54140.7.1_25-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420066 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1213982 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213982 | - | - |
