Assay Details
| Gene Symbol: Mfsd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027122.3 | NP_081398.2 | 4 - 5 | 1191 | 63 | |
| GenBank mRNA | AK009269.1 | - | 4 - 5 | 1270 | 63 | |
| AK089539.1 | - | 4 - 5 | 1192 | 63 | ||
| BC019171.1 | - | 4 - 5 | 1218 | 5003354 | 63 | |
| BC085305.1 | - | 4 - 5 | 1255 | 6399056 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350057 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350055 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350058 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P528463 | AK089539.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P528463 | BC019171.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P528463 | NM_027122.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P528463 | AK009269.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P528463 | AK089539.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P528463 | BC019171.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P528463 | NM_027122.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P528463 | AK009269.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 1645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970717 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P528463 | AK089539.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P528463 | BC019171.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P528463 | NM_027122.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P528463 | AK009269.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P528463 | AK089539.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P528463 | BC019171.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P528463 | NM_027122.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P528463 | AK009269.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350046 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08423151 | - | - |
| Whole Mouse Genome G4122A | A_52_P528463 | AK089539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P528463 | BC019171.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P528463 | NM_027122.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P528463 | AK009269.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P528463 | AK089539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P528463 | BC019171.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P528463 | NM_027122.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P528463 | AK009269.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117098 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12362768 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12362701 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12107290 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12107277 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12107282 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12107280 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12362751 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350057 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350058 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12107280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12362721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12183631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350050 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10424993 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430073 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424999 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430074 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424994 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424996 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424997 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424998 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451433_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10424991 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430071 | - | - |
| Mouse Genome 430 2.0 Array | 1451433_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451433_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69572_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69572_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500004429.mm.1 | NM_027122.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004429.mm.1 | NM_027122.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004429.mm.1 | TC1500000606.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004430.mm.1 | NM_027122.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004430.mm.1 | NM_027122.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004430.mm.1 | TC1500000606.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004432.mm.1 | NM_027122.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004432.mm.1 | NM_027122.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004432.mm.1 | TC1500000606.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004431.mm.1 | NM_027122.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004431.mm.1 | NM_027122.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004431.mm.1 | TC1500000606.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451433_at | - | - |
| Mouse Expression Set 430 | 1451433_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451433_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451433_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451433_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451433_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 320727 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4780670 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 510524 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000510524 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004780670 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2742136 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2644480 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2644480 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2742136 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002574.1_42-S | - | - |
| mouse-6 v1.1 (Illumina) | scl47786.6.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 510524 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4780670 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47786.6.1_30-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002574.1_42-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780670 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510524 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2742136 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2644477 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2644480 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2742136 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2644477 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2644480 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl47786.6.1_30-S | - | - |

Mfsd3
Genomic Map