Assay Details
Gene Symbol: Hfe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027126.4 | NP_081402.3 | 4 | 1310 | 69 | |
GenBank mRNA | AJ557515.1 | - | 4 | 1310 | 69 | |
AK009636.1 | - | 1 | 557 | 69 | ||
AK014082.1 | - | 4 | 1283 | 69 | ||
BC022603.1 | - | 4 | 1334 | 69 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206824 | AK014082.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P206824 | AK014082.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211530 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590287 | AK141786.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590287 | BC034586.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590287 | BC042467.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590287 | BC058763.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590287 | NM_013415.5 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590287 | X16645.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 948147 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122554 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130764 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206824 | AK014082.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P206824 | AK014082.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206824 | AK014082.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206824 | AK014082.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349760 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349743 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349772 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349758 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349744 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349755 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349732 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349753 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349730 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349770 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22349757 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130764 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04997890 | - | - |
Whole Mouse Genome G4122A | A_51_P206824 | AK014082.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P206824 | AK014082.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115353 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130764 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02872148 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02872147 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02872140 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02872142 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02872145 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02872146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22349770 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02749513 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03069717 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03192951 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349733 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349746 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03192948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349773 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349792 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349771 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349729 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349755 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349738 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349796 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349770 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349801 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349725 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349782 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02357437 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02469420 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349730 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03192964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02872142 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349763 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349724 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02289170 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349793 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349794 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349767 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02872146 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03192953 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02636289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349728 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349788 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02469422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02871310 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349737 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22349785 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10494425 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10494424 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10494427 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10494426 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429223_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1454137_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10494423 | - | - |
Mouse Genome 430 2.0 Array | 1454137_s_at | - | - |
Mouse Genome 430 2.0 Array | 1429223_a_at | - | - |
Mouse Genome 430 2.0 Array | 1429223_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1454137_PM_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69585_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69585_PM_at | - | - |
Mouse Expression Set 430 | 1429223_PM_a_at | - | - |
Mouse Expression Set 430 | 1429223_a_at | - | - |
Mouse Expression Set 430 | 1454137_s_at | - | - |
Mouse Expression Set 430 | 1454137_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454137_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454137_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429223_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429223_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3850148 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3850148 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2100138 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003850148 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002100138 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250363 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2736987 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1250363 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2736987 | - | - |
mouse-6 v1.1 (Illumina) | scl0001933.1_4-S | - | - |
mouse-6 v1.1 (Illumina) | scl22860.4.705_3-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2100138 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3850148 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl22860.4.705_3-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001933.1_4-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850148 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100138 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2688089 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2688092 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688089 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688092 | - | - |
