Assay Details
| Gene Symbol: Gpx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027127.2 | NP_081403.1 | 1 - 2 | 218 | 62 | |
| GenBank mRNA | AK009378.1 | - | 1 - 2 | 219 | 62 | |
| BC019664.1 | - | 1 - 2 | 217 | 3988837 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpx8
Genomic Map