Assay Details
Gene Symbol: Mif4gd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243584.1 | NP_001230513.1 | 4 - 5 | 587 | 77 | |
NM_027162.4 | NP_081438.4 | 3 - 4 | 871 | 77 | ||
GenBank mRNA | AK003708.1 | - | 648 | 77 | ||
AK010175.1 | - | 4 - 5 | 541 | 77 | ||
AK150749.1 | - | 4 - 5 | 541 | 77 | ||
BC019515.1 | - | 4 - 5 | 498 | 4460171 | 77 | |
BC026740.1 | - | 4 - 5 | 547 | 4015105 | 77 | |
BC055812.1 | - | 3 - 4 | 871 | 4934728 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
